missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ Acid-Treated Low-Metal TCLP Filters
Binder free filters
Supplier: Cytiva 1810125
Description
Binder free filters
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm
- Resistant to acids and bases
- Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
- Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs
Compliance
Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.
Specifications
GEâs Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action. | |
Borosilicate Glass | |
Determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources | |
Circle | |
TCLP Limits Test Filter | |
4.92 in. |
Disc | |
0.7 μm | |
50 pieces | |
420 μm | |
60 mL/min | |
125 mm |