Please login to your online account to display your discounted pricing

Whatman™ Acid-Treated Low-Metal TCLP Filters


Binder free filters

Manufacturer: whatman™  1810047

 View more versions of this product

Catalog No. 09-921-7

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Diameter (English) Outside 1.8 in.
Particle Retention 0.7μm
Diameter (Metric) Outside 4.7cm
For Use With (Application) Determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources
Material Borosilicate Glass
Accessories

Accessories for Catalog Number: 099217

Item Mfr. No. Item Description Price Per Unit Quantity  
09-753-50
Fisherbrand™ Filter Forceps
fisherbrand™
SDI25510
Easy, effortless handling of membranes and glass fiber filters Each for $11.07