Please login to your online account to display your discounted pricing
Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters
Handle multiple samples at one time
Manufacturer: whatman™  WHTCR0000006
 View more versions of this product
 Alert
The six position multicompressor(#09-927-13) is only for use with plastic MiniUniprep.
The eight position multicompressor (#09-924-491) is only for use with MiniUniprep G2 glass.
Catalog No. 09-927-13
  • / Each
Description & Specifications
Specifications
Housing Material Polypropylene
Type Multi-sample compressor, Six positions
Material PTFE Coated Silicone Rubber
Applications Mini-UniPrep G2 syringeless filters