Please login to your online account to display your discounted pricing
Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters
Handle multiple samples at one time
$30.80 - $669.70
Specifications
Applications Mini-UniPrep G2 syringeless filters
View More Specs
 Alert
The six position multicompressor(#09-927-13) is only for use with plastic MiniUniprep.
The eight position multicompressor (#09-924-491) is only for use with MiniUniprep G2 glass.
Products
Catalog Number Mfr. No. Type Price Quantity    
09-927-13
whatman™
WHTCR0000006
Multi-sample compressor, Six positions Each for $621.27
09-924-490
whatman™
MUPG2HCPWC1
Hand compressor, One position Each for $30.80
09-924-491
whatman™
MUPG2MCPWC8
Multi-sample compressor, Eight positions Each for $669.70
09-924-492
whatman™
MUPG2MCWT8
Eight Vial Compressor Tray Each for $45.27
Description & Specifications
Specifications
Applications Mini-UniPrep G2 syringeless filters
Compressors
  • Handheld devices to aid in compression of Mini-UniPrep and Mini-UniPrep G2 filter units
  • Compressor Tray
    • Speeds up the sample prep process
    • Extra trays allow pre-loading of samples
    • For use with 8-position multi compressor (#09-924-491)