Please login to your online account to display your discounted pricing

Whatman™ 96-Well Polypropylene UniPlate™ Microplates

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ 96-Well Polypropylene UniPlate™ Microplates withstand temperatures to -80°C for long-term storage.

$322.62 - $322.62

Specifications

Color Natural
For Use With (Application) For homogeneous assays techniques utilized in high-throughput screening
Material Polypropylene
No. of Wells 96
View More Specs
Products
Catalog Number Mfr. No. Bottom Volume (Metric) Well Well Design Price Quantity    

09-003-36

 
whatman™
77015200
Pack of 25 Each for $322.62
Description & Specifications

Specifications

Color Natural
For Use With (Application) For homogeneous assays techniques utilized in high-throughput screening
Material Polypropylene
No. of Wells 96

 

  • One-piece molded collection and analysis plates
  • Conforms to ANSI/SBS microplate standard
  • Prevent optical cross talk in light emitting assays
  • Untreated
  • Ideal for filtrate collection, homogeneous assay techniques used in high-throughput screening (HTS)

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.