Please login to your online account to display your discounted pricing
Whatman™ 4mm Puradisc™ Nonsterile PVDF Syringe Filters
Designed for high tensile strength, high solvent resistance, and low protein binding. Whatman™ 4mm Puradisc™ Nonsterile PVDF Syringe Filters are well-suited for biomedical filtration, sterilization filtration, and HPLC sample preparation.
$288.70 - $1,861.41
Specifications
Material Polyvinylidene Difluoride (PVDF)
Housing Material Pigment-free Polypropylene
Diameter (Metric) 4mm
Pressure Rating 5.2bar
Inlet Style Female Luer Lock
View More Specs
Products
Catalog Number Mfr. No. Outlet Style Pore Size Price Quantity    
09-302-108
whatman™
WHT67920402*
Male Luer 0.2μm Pack of 50 Each for $1,788.24
09-910-1
whatman™
WHT67770402
Tube tip 0.2μm Pack of 50 Each for $288.70
09-910-11
whatman™
WHT67790402
Male Luer 0.2μm Pack of 100 Each for $409.63
09-910-12
whatman™
WHT67790404
Male Luer 0.45μm Pack of 100 Each for $409.63
09-926-38
whatman™
WHT67920404
Male Luer 0.45μm Pack of 500 Each for $1,861.41
09-926-40
whatman™
WHT67770404
Tube tip 0.45μm Pack of 50 Each for $289.52
Description & Specifications
Specifications
Material Polyvinylidene Difluoride (PVDF)
Housing Material Pigment-free Polypropylene
Diameter (Metric) 4mm
Pressure Rating 5.2bar
Inlet Style Female Luer Lock
Volume (Metric) Up to 2mL
Filtration Area (Metric) 0.2cm2
  • PVDF filtration media
  • Good resistance to organic solvents
  • Nonsterile
  • Polypropylene housing
  • Available with male Luer or tube-tip outlet  (allows direct filtration into microvial inserts)
  • Female Luer lock inlet
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.