Please login to your online account to display your discounted pricing
Whatman™ 13mm Puradisc™ Nonsterile PVDF Syringe Filters
Designed for high tensile strength, high solvent resistance, and low protein binding. Whatman™ 13mm Puradisc™ Nonsterile PVDF Syringe Filters are well-suited for biomedical filtration, sterilization filtration, and HPLC sample preparation.
$303.89 - $4,522.02
Specifications
Diameter (Metric) 13mm
Type Syringe Filter
Housing Material Polypropylene
Filtration Area (Metric) 1.3cm2
Material Polyvinylidene Difluoride (PVDF)
View More Specs
Products
Catalog Number Mfr. No. Outlet Style Pore Size Price Quantity    
05-713-409
whatman™
6765-1304
Male Luer 0.45μm Pack of 2000 Each for $4,522.02
09-302-113
whatman™
67651302
Male Luer 0.2μm Pack of 2000 Each for $4,479.22
09-910-14
whatman™
67791304
Male Luer 0.45μm Pack of 100 Each for $450.70
09-910-2
whatman™
67771302
Tube Tip 0.2μm Pack of 50 Each for $303.89
09-910-22
whatman™
67921302
Male Luer 0.2μm Pack of 500 Each for $1,913.52
09-910-3
whatman™
67771304
Tube Tip 0.45μm Pack of 50 Each for $303.89
09-910-23
whatman™
67921304
Male Luer 0.45μm Pack of 500 Each for $1,913.52
09-910-13
whatman™
6779-1302
Male Luer 0.2μm Pack of 100 Each for $443.83
Description & Specifications
Specifications
Diameter (Metric) 13mm
Type Syringe Filter
Housing Material Polypropylene
Filtration Area (Metric) 1.3cm2
Material Polyvinylidene Difluoride (PVDF)
Pressure Rating 5.2bar
Inlet Style Female Luer lock
Volume (Metric) Up to 10mL
Hold up Volume (After Air Purge) <25μL
  • PVDF filtration media
  • Good resistance to organic solvents
  • Nonsterile
  • Polypropylene housing
  • Autoclavable available
  • Available with male Luer slip or tube-tip outlet (allows direct filtration into microvial inserts)
  • Inlet is female Luer lock type
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.