missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wells Lamont™ Industrial Extra-Heavyweight Terry Cloth Gloves

Made from high-loop terry cloth, a natural insulator

$314.92 - $314.92

Specifications

Color Brown,White
Material Terrycloth
Size X-Large
For Use With (Application) flexibility and comfort in most applications
Glove Cuff Style Gauntlet
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Temperature (English) Max. Temperature (Metric) Max Price Quantity  
Catalog Number Mfr. No. Temperature (English) Max. Temperature (Metric) Max Price Quantity  
19-013299 Jomac Canada
636HRL
450°F 232.22°C Dozen of 12 Pair for $314.92
Case of 5 Dozen for $1,574.60
Specifications

Description

  • Withstand temperatures up to 450°F (232°C )
  • Maintain flexibility and comfort in most applications
  • An excellent alternative to asbestos or leather
  • Lined, preshrunk, and launderable
  • Gauntlet cuff: 5.5 in. long (14cm)
  • Colors: Brown and White

Description & Specifications

Specifications

Brown,White
X-Large
Gauntlet
Terrycloth
flexibility and comfort in most applications
Ambidextrous