Welch™ Replacement Element for Vacuum Pumps
Accessory for Welch vaccum pumps
Manufacturer: Welch™ 1420E02
Description
- For acid neutralization trap and oil mist elinimator
- Changes from white to bluish-transparent when spent
Specifications
Model 1400N vacuum pump | |
Acid Neutralization Trap Element |