Please login to your online account to display your discounted pricing
Welch™ Oil-Mist Eliminator for Vacuum Pumps
Accessories for vacuum pumps
$452.94 - $1,661.37
Specifications
For Use With (Application) Vacuum Pumps
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity    
01-108-1A
welch™
1416E01
Model 1400N vacuum pump Pump Adaptor Kit Each for $452.94
01-108A
welch™
1416D
Model 1400N, 1402N, 1376N vacuum pump Oil Mist Eliminator Each for $1,661.37
Description & Specifications
Specifications
For Use With (Application) Vacuum Pumps
Oil-Mist Eliminator
  • Hermetically sealed
  • SS w/borosilicate microfiber element, flouroelastomer gaskets
  • Prevents corrosive exhaust gases from escaping
  • Accepts 13/16 in. hose
  • 12.7W x 19.68 in. H
Pump-Adapter Kit
  • For use with Oil-Mist Eliminator