Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
$421.80 - $1060.37
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
View More Specs
Products 3
Catalog Number Mfr. No. Pump Type Price Quantity  
Catalog Number Mfr. No. Pump Type Price Quantity  
01-258-119
View Documents Promotion Details
Welch™
1475K10
Hand Held Pipette
Each for $1,060.37
01-258-120
View Documents Promotion Details
Welch™
1475K20
Spare Receiver Kit
Each for $649.91
01-258-121
View Documents Promotion Details
Welch™
1475K21
Replacement Hydrophobic Filter
Each for $421.80
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station
SDS
missing translation for 'documents'
missing translation for 'documents'