Please login to your online account to display your discounted pricing

American Educational Products Weather Map/Plotting Chart


Double-sided chart helps students record and forecast weather conditions.

Manufacturer: american educational products  HUB464

 View more versions of this product

Catalog No. S45136-1

  • / Each
 

Description & Specifications

Specifications

Product Type Weather Climagraph
For Use With (Application) Help students record and forecast weather conditions
Classification Science
Includes Locations of reporting cities and world map outline on the reverse side, two climagraphs for plotting comparative temperatures and precipitation