missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

American Educational Products Weather Map/Plotting Chart

Double-sided chart helps students record and forecast weather conditions.

$22.34 - $243.11

Specifications

Classification Science
For Use With (Application) Help students record and forecast weather conditions
Includes Locations of reporting cities and world map outline on the reverse side, two climagraphs for plotting comparative temperatures and precipitation
View More Specs

Includes: Study guide

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S45136-1 View Documents American Educational Products
HUB464
Weather Climagraph Each for $22.34
 
S45523 View Documents American Educational Products
HUB462
Weather Map/Plotting Chart Each for $243.11
 
Description

Description

  • U.S. map side includes major cities, weather forecast table, and symbol explanation
  • World map side includes two climagraphs for comparing temperature and precipitation
  • L × W: 44 × 50 in. (112 × 127cm)

Order Info

Corresponding worksheets sold in pads of 50 sheets.

Specifications

Specifications

Science
Help students record and forecast weather conditions
Locations of reporting cities and world map outline on the reverse side, two climagraphs for plotting comparative temperatures and precipitation