Please login to your online account to display your discounted pricing

Wearwell™ PermaTack™ Contamination Control Mats

A high-performance, industrial strength contamination control mat

$971.81 - $1,400.93

Specifications

Color Blue
For Use With (Application) Remove particles off shoes or wheels
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity    

19-151-142

 
wearwell co.
092.18X27X48BL
68.5 x 122cm (27 x 48 in.) Each for $971.81

19-151-144

 
wearwell co.
092.18X36X54BL
91.4 x 137cm (36 x 54 in.) Each for $1,400.93
Description & Specifications

Specifications

Color Blue
For Use With (Application) Remove particles off shoes or wheels

  • Permanently tacky surface removes even the finest particles off shoes or wheels
  • 98% to 100% of contamination will be removed if each foot touches the mat three times
  • Anti-microbial blue vinyl polymer
  • High performance backing provides structural durability
  • Unique adhesive backing bonds to the floor

One year.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.