Please login to your online account to display your discounted pricing

Tocris™ WAY 629 hydrochloride

Selective 5-HT2C agonist

$181.19 - $762.58

Specifications

Product Type WAY 629 hydrochloride
Inhibitors WAY 629 hydrochloride
Molecular Weight 262.78
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

217310

 
tocris™
217310
10mg Each for $181.19

217350

 
tocris™
217350
50mg Each for $762.58
Description & Specifications

Specifications

Product Type WAY 629 hydrochloride
Inhibitors WAY 629 hydrochloride
Molecular Weight 262.78
Purity >98

Selective 5-HT2C agonist. Ki values are 56, 2350, 1575 and 815nM for human recombinant 5-HT2C, 5-HT2A, 5-HT6 and 5-HT7 receptors respectively. Stimulates intracellular Ca2+ mobilization in CHO cells expressing human 5-HT2C receptors (EC50 = 72nM). Reduces food intake in rats.

  • Chemical Name: 1,2,3,4,8,9,10,11-Octahydro[1,4]diazepino[6,7,1-jk]carbazole hydrochloride
  • Formula: C15H18N2.HCl
  • CAS Number: 57756-44-2
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98