Please login to your online account to display your discounted pricing

WAY 213613, Tocris

Potent, non-substrate EAAT2 inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material WAY 213613
Assay Percent Range >99%
Molecular Formula C16H13BrF2N2O4
Formula Weight 415.19
Synonym N-[4-(2-Bromo-4, 5-difluorophenoxy)phenyl]-L-asparagine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

265250

 
r & d systems
RDS265250
50mg Each for $1,073.10

265210

 
r & d systems
RDS265210
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 415.19
Quantity 50mg
Chemical Name or Material WAY 213613
Assay Percent Range >99%
Molecular Formula C16H13BrF2N2O4
CAS 868359-05-1
Synonym N-[4-(2-Bromo-4, 5-difluorophenoxy)phenyl]-L-asparagine
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature