missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, ultrapure, Spectrophotometric Grade, Alfa Aesar™

Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5μ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T.

Manufacturer:  Alfa Aesar 19391K7

 View more versions of this product

Catalog No. AA19391K7



Specifications

Specifications

Water
Colorless liquid
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Sprectrophotometric
Colorless
7732-18-5
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Liquid
23 hPa (17mm Hg) at 20°C
4L

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
SDS Documents