missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, ultrapure, Spectrophotometric Grade, Alfa Aesar™
Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5μ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T.
$63.04 - $200.14
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen monoxide, dihydrogen oxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen monoxide, dihydrogen oxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
H2O | |
dihydrogen monoxide, dihydrogen oxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Spectrophotometric | |
0°C | |
Colorless | |
1L | |
Water |
Colorless liquid | |
MFCD00011332 | |
Miscible with methanol,acetone,ethanol. | |
O | |
18.015 | |
CHEBI:15377 | |
Liquid | |
23 hPa (17mm Hg) at 20°C | |
1 | |
100°C | |
1.333 |
Safety and Handling
EINECSNumber : 231-791-2
TSCA : Yes
Recommended Storage : Ambient temperatures