missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Manufacturer:  Thermo Scientific™ J70783XCR

 View more versions of this product

Catalog No. AAJ70783XCR



Specifications

Specifications

Water
H2O
dihydrogen monoxide
O
18.015
CHEBI:15377
Liquid
Nalgene bottle
100°C
1.000
7732-18-5
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Molecular Biology
0°C
10 x 1mL

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen monoxide
CHEBI:15377
O
Safety and Handling

Safety and Handling

TSCA : Y

Documents