Please login to your online account to display your discounted pricing

Alfa Aesar™ Water, RNase, DNase-free

Chemical Identifiers

Chemical Name or Material Water
CAS 7732-18-5
Name Note RNase, DNase-free
Formula Weight 18.02
MDL Number MFCD00011332
Beilstein 2050024
Merck Index 14,10039
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60610EQC

  SDS
alfa aesar™
J60610EQC
3.8L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 18.02
Chemical Name or Material Water
TSCA TSCA
Physical Form Liquid
CAS 7732-18-5
Density 1.000g/mL
Color Clear colorless
Boiling Point 100°C
Quantity 3.8L
Name Note RNase, DNase-free
Beilstein 2050024
Merck Index 14,10039
Odor Odorless
RTECS Number ZC0110000
MDL Number MFCD00011332

Applications
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.

Solubility
It is soluble in water.

Notes
Toxic agents such as DEPC are not used in purification procedures.