LaMotte™ Water Quality Educator and Monitoring Outfit
Ideal for long-term water-quality monitoring.
Manufacturer:  LaMotte™ R5870
 View more versions of this product
Catalog No. S66224
Includes:
  • CD-ROM benchmarks and compares data for each test factor, explains typical causes and effects of higher and lower quality levels
  • The Monitor's Handbook (71 pages)
  • 100 tests for pH
  • 50 tests for the following five factors: nitrate-nitrogen, phosphate, dissolved oxygen, total alkalinity, turbidity, unlimited temperature tests, aquatic macroinvertebrate bioassessment
  • Lesson plans for grades 5 to 8 and 9 to 12
Description
Description
  • Tests for seven basic water-quality test factors Prepares students for field tests or classroom work
  • Set up a water-quality monitoring program, procure measurements, and interpret results
  • Soft-sided tote made of durable fabric, solid rubber bottom ready for field work; handy side pockets hold thermometer, pens, writing tablets and other tools
Order Info
System requirements:
  • Windows™: 64MB available RAM, Windows 98, 2000, XP™ or Vista™
  • Macintosh™: 128MB available RAM, OSX™ 10.1.5, 10.2.6 or 10.3
Specifications
Specifications
Long term water quality monitoring
Set of refill reagents as originally supplied in the complete kit
Reagent Refill