LaMotte™ Water Quality Classroom Studies
Reinforce key concepts on water treatment and environmental studies.
Manufacturer:  LaMotte™ 5967
 View more versions of this product
Catalog No. S66988
Description
Description
  • Teacher's manual has tips for organizing the demonstration, test factor information and suggestions for additional experiments
  • Also include introductory information, reproducible student procedures/data sheets, lesson plan and reagent composition information
Dissolved Oxygen Lab: Discover the relationship between temperature and oxygen saturation.
Nutrients in Water Lab: Nitrate and phosphate have a key role in indicating pollution. Prepared samples include levels of these nutrients for measuring and discussion.
Tapwater Hardness Lab: Provided sample demonstrates the quantitative and aesthetic qualities of drinking water.
Water Treatment Lab: Investigates the water treatment process by examining the odor, color and turbidity of drinking water.
Order Info
Grades 5 to 8. Class size: 25 demonstrations.
Specifications
Specifications
Water quality studies
5 to 8
25 Demonstrations
Tapwater Hardness Lab