Please login to your online account to display your discounted pricing

LaMotte™ Water Quality Classroom Studies


Reinforce key concepts on water treatment and environmental studies.

Manufacturer: lamotte™  5963

 View more versions of this product

Catalog No. S66983

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Water quality studies
Grade 5 to 8
Product Type Dissolved Oxygen Lab
Sufficient For 25 demonstrations