Please login to your online account to display your discounted pricing
LaMotte™ Water Pollution Detection Demonstration Outfit
Use overhead projector to demonstrate tests.
Manufacturer: lamotte™  LAMR5905
 View all options for this product
Catalog No. S45075A
Description & Specifications
For Use With (Application) To introduce concepts of water pollution detection
Product Type Reagent Refill
Introduce the basic concepts of detecting water pollution by field test procedures and overhead projections. You can demonstrate testing for the following: alkalinity, ammonia, calcium, carbon dioxide, chlorides, chlorine, chromium, copper, cyanide, dissolved solids, hardness, iron, magnesium, nitrates, pH, phosphates, salinity, and sulfides.
The two texts included (Our Environment Battles Water Pollution and Investigating Water Problems) provide useful background information on water pollution and testing.