Please login to your online account to display your discounted pricing

LaMotte™ Water Pollution Detection Demonstration Outfit


Use overhead projector to demonstrate tests.

Manufacturer: lamotte™  LAM5905

 View more versions of this product

Catalog No. S45075

  • / Each
 

Description & Specifications

Specifications

No. of Tests 20.0
Grade 5+
For Use With (Application) To introduce concepts of water pollution detection
Product Type Water Pollution Introductory Kit