missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Optima™ LC/MS Grade, Fisher Chemical™

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.

Supplier:  Thermo Fisher Scientific W6500

 View more versions of this product

Catalog No. W6500




Description

Description

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.

Specifications

Specifications

≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
7
Colorless
100°C
H2O
15, 10237
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Glass Bottle
17.5mmHg at 20°C
1.000g/cm³
500 mL
Water
7732-18-5
0°C
LC-MS & UHPLC-UV
11332
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
OPTIMA™ LC-MS
≤1ppm
0.03 micron filtered.
Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
11332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention.

NFPA

Health: 0
Flammability: 0
Instability: 0

SDS Documents

Documents

Promotions

Promotions