missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 327390050

 View more versions of this product

Catalog No. AC327390050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

18.02
5L
15, 10237
Plastic Jerrycan
1.0000g/mL
0.5ppm max.
Water
DNase, RNase, Protease free
1ppb max.
< 0.0001% (at bottling)
MFCD00011332
Liquid
Clear
16,383
For molecular biology
>18 megaohm (at time of manufacture)
none detected
H2O
0.1ppm max.
TOC: < 50ppb
7732-18-5
H2O

Chemical Identifiers

7732-18-5
H2O
MFCD00011332
For molecular biology
H2O
18.02
Water, DNase, RNase, Protease free

Safety and Handling

SDS & Safety Documents