Please login to your online account to display your discounted pricing

Water, Microbial Cell Culture Grade, Fisher BioReagents

$34.38 - $56.36

Chemical Identifiers

Chemical Name or Material Water
Name Note 0.03μm filtered and autoclaved
CAS 7732-18-5
Purity Grade Microbial Cell Culture
Molecular Formula H2O
Formula Weight 18.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2820-100

 
fisher scientific company
BP2820100
Nalgene Poly Bottle 100mL Each for $34.38
Case of 6 Each for $206.28

BP2820-500

 
fisher scientific company
BP2820500
Nalgene Poly Bottle 500mL Each for $42.00
Case of 6 Each for $252.00

BP2820-1

 
fisher scientific company
BP28201
Nalgene Poly Bottle 1L Each for $56.36
Case of 6 Each for $338.16
Description & Specifications

Specifications

Boiling Point 100°C
Formula Weight 18.01
Quantity 100mL
Density 1.000g/cm³
Color Clear Colorless
Conductivity 2μS/cm Max.
Purity Grade Microbial Cell Culture
Physical Form Liquid
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.03 Micron filtered and autoclaved
Barium (Ba) 10ppb max.
Calcium (Ca) 20 ppb max.
Cadmium (Cd) 10 ppb max.
Aluminum (Al) 10ppb max.
Iron (Fe) 10 ppb max.
Potassium (K) 10 ppb max.
Resistivity 16 MEGOHM-cm min.
Selenium (Se) 10ppb max.
Sodium (Na) 20ppb max.
Arsenic (As) 10ppb max.
Chemical Name or Material Water
Chromium (Cr) 10 ppb max.
Name Note 0.03μm filtered and autoclaved
Copper (Cu) 10 ppb max.
Molecular Formula H2O
CAS 7732-18-5
ChemAlert Storage Symbol Gray
Lead (Pb) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Health Hazard 2 Low hazard for usual industrial or commercial handling.
Nickel (Ni) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Health Hazard 3 Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0
Additional Information Vapor Pressure: 17.5mmHg at 20°C
Tin (Sn) 10ppb max.
Yeast and Mold < 1 CFU/mL
Cobalt (Co) 10 ppb max.
Endotoxin 0.025 EU/mL max.
Growth Promotion Pass Test
Manganese (Mn) 10 ppb max.
Oxidizable Substances Pass Test
Packaging Nalgene Poly Bottle
Silver (Ag) 10ppb max.
Vanadium (V) 10ppb max.
Bacteria < 1 CFU/mL

Ultrapure water for preparing microbiological growth media.

Safety and Handling
 

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0

Low hazard for usual industrial or commercial handling.