missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, LC/MS grade, Alfa Aesar™

Used as mobile phase solvent in LC-MS analysis

$265.26 - $505.11

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA47146M6
View Documents
Thermo Fisher Scientific
47146M6
4 x 4L
Case of 4 Each for $505.11
 
AA47146Y0
View Documents
Thermo Fisher Scientific
47146Y0
6 x 1L
Case of 6 Each for $265.26
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
CHEBI:15377
O

Specifications

7732-18-5
MFCD00011332
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
LC-MS
0°C
Colorless
7 (at 20°C)
1.34
H2O
2050024
Soluble in water; Insoluble in polar liquids.
O
18.015
CHEBI:15377
Liquid
Odorless
1 g/cm3
100°C
4 x 4L
Water
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

RTECSNumber : ZC0110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS