missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, for HPLC gradient grade, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 268300040
Description
For HPLCSpecifications
Water | |
7732-18-5 | |
Vapor Pressure: 17.5mmHg at 20°C | |
H2O | |
Yes | |
16,383 | |
2mL/min | |
dihydrogen monoxide, dihydrogen oxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
0.002 A.U. max. | |
0.0005% max. | |
acetonitrile | |
0.89 mPa.s (25°C) | |
Colorless | |
4L | |
1 |
For HPLC gradient grade | |
0.04 A.U.F.S. max. | |
H2O | |
MFCD00011332 | |
C18 reversed phase | |
15, 10237 | |
lineair from 100% A to 100% B over 20 minutes | |
: | |
O | |
18.015 | |
CHEBI:15377 | |
HPLC Gradient | |
0.0003% max. | |
water | |
254nm | |
1.0000g/mL | |
100.0°C | |
1.3334 |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen monoxide, dihydrogen oxide | |
CHEBI:15377 | |
O |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000
TSCA : TSCA