missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, HPLC Grade, Alfa Aesar™, Ultrapure

Manufacturer:  Alfa Aesar 22934M6

 View more versions of this product

Catalog No. AA22934M6



Specifications

Specifications

Water
HPLC
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
1.000
7732-18-5
H2O
pure water density standard ukas is/iec17025 and is guide 34 certified density: 0.9982 g/ml at 20 c density: 0.9970 g/ml at 25 c, bacteristatic water fr injectin in plastic cntainer, tris-1 10-phenanthrline irn ii sulfate slutin, water distilled cnductivity r f similar purity, sterile water fr injectin in plastic cntainer, water ultrapure spectrphtmetric grade, buffer slutin ph 10.00 clr-cded blue, water fr injectin in cntainers jp17, water fr uhplc fr mass spectrmetry, water with trifluracetic acid
O
18.015
CHEBI:15377
Liquid
4 x 4L

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
pure water density standard ukas is/iec17025 and is guide 34 certified density: 0.9982 g/ml at 20 c density: 0.9970 g/ml at 25 c, bacteristatic water fr injectin in plastic cntainer, tris-1 10-phenanthrline irn ii sulfate slutin, water distilled cnductivity r f similar purity, sterile water fr injectin in plastic cntainer, water ultrapure spectrphtmetric grade, buffer slutin ph 10.00 clr-cded blue, water fr injectin in cntainers jp17, water fr uhplc fr mass spectrmetry, water with trifluracetic acid
CHEBI:15377
O
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

SDS Documents