missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, HPLC Grade, Alfa Aesar™, Ultrapure

$61.30 - $370.09

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen monoxide, dihydrogen oxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA22934K2
View Documents
Thermo Fisher Scientific
22934K2
1L
Each for $61.30
 
AA22934K7
View Documents
Thermo Fisher Scientific
22934K7
4L
Each for $133.11
 
AA22934M6
View Documents
Thermo Fisher Scientific
22934M6
4 x 4L
Each for $370.09
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen monoxide, dihydrogen oxide
CHEBI:15377
O

Specifications

7732-18-5
MFCD00011332
Miscible with methanol,acetone,ethanol.
O
18.015
CHEBI:15377
Liquid
0°C
100°C
1.333
H2O
dihydrogen monoxide, dihydrogen oxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
HPLC
1.000
1L
Water
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS