missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water Hardness Buffer, without Magnesium EDTA, Ricca Chemical

$89.27 - $89.27

Chemical Identifiers

CAS 7732-18-5,1336-21-6,12125-02-9
CAS Min % 62.0,31.88,4.19
CAS Max % 64.0,33.18,4.36
Physical Form Liquid
Chemical Name or Material Water Hardness Buffer, Without Magnesium EDTA
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
9201-32 Ricca Chemical Company
RI920132
Poly natural 1L Each for $89.27
9201-16 Ricca Chemical Company
RI920116
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1336-21-6,12125-02-9
64.0,33.18,4.36
Water Hardness Buffer, Without Magnesium EDTA
62.0,31.88,4.19
Liquid

Specifications

7732-18-5,1336-21-6,12125-02-9
64.0,33.18,4.36
Poly natural
1L
62.0,31.88,4.19
Liquid
Clear colorless
Water Hardness Buffer, Without Magnesium EDTA