Please login to your online account to display your discounted pricing

Water Hardness Buffer, Odorless, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR920500020F

 View more versions of this product

Catalog No. R920500020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Colorless to slightly yellow
Physical Form Liquid
Quantity 20L
Chemical Name or Material Water Hardness Buffer
CAS Max % 4.81
CAS 6381-92-6
CAS Max % 0.35
CAS 7791-18-6
CAS Min % 0.18
CAS Max % 0.19
CAS 7732-18-5
CAS Min % 71.0
CAS Max % 74.0
CAS 141-43-5
Packaging Cubitainer
CAS Min % 21.62
CAS Max % 22.5
CAS 7647-01-0
CAS Min % 4.62
CAS Min % 0.34
Name Note Odorless

Specifications

Chemical Name or Material Water Hardness Buffer
Name Note Odorless
CAS 7732-18-5
CAS Min % 71.0
CAS Max % 74.0
CAS 141-43-5
CAS Min % 21.62
CAS Max % 22.5
CAS 7647-01-0
CAS Min % 4.62
CAS Max % 4.81
CAS 6381-92-6
CAS Min % 0.34
CAS Max % 0.35
CAS 7791-18-6
CAS Min % 0.18
CAS Max % 0.19
Physical Form Liquid