Please login to your online account to display your discounted pricing

Water Hardness Buffer, with Magnesium, Certified, LabChem™

$52.14 - $231.86

Chemical Identifiers

Chemical Name or Material Water Hardness Buffer
CAS 7732-18-5
CAS Max % 88.1
CAS 12125-02-9
CAS Max % 6.8
CAS 1336-21-6
CAS Max % 4.6
CAS 14402-88-1
CAS Max % 0.5
Name Note with Magnesium
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC268101

 
lab chem
LC268101
Poly Bottle 500mL Each for $52.14

LC268102

 
lab chem
LC268102
Poly Bottle 1L Each for $80.71

LC268104

 
lab chem
LC268104
Poly Bottle 4L Each for $231.86
Description & Specifications

Specifications

Formula Weight 35.05
Purity Grade Certified Grade
CAS Max % 88.1
UN Number UN2672
CAS 1336-21-6
CAS Max % 0.5
Linear Formula NH4OH
Recommended Storage Room Temperature
Identification Passes Test
CAS 14402-88-1
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.
Specific Gravity 0.96g/mL
Color Clear, colorless
CAS 7732-18-5
CAS 12125-02-9
Solubility Soluble in water
Quantity 500mL
Packaging Poly Bottle
CAS Max % 6.8
Physical Form Liquid
Suitability Passes Test
Health Hazard 1 Danger
Density 0.96g/mL
Chemical Name or Material Water Hardness Buffer
Health Hazard 3 GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
CAS Max % 4.6
Name Note with Magnesium
Safety and Handling

Danger

GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.

Room Temperature