missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water Hardness Buffer, with Magnesium, Certified, LabChem™

Supplier:  LabChem LC268102

 View more versions of this product

Catalog No. LC268102



Description

Description

Specifications

Specifications

Water Hardness Buffer
88.1,6.8,4.6,0.5
12125-02-9,1336-21-6,14402-88-1
0.96g/mL
1 L
NH4OH
UN2672
Soluble in water
Certified
with Magnesium
Colorless
Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Liquid
Passes Test
Poly Bottle
0.96g/mL
35.05
SDS
Documents

Documents

Promotions

Promotions