Please login to your online account to display your discounted pricing

Water Hardness Buffer, LabChem™

$64.91 - $87.26

Chemical Identifiers

Chemical Name or Material Water Hardness Buffer
CAS 7732-18-5
CAS Max % 93.25
CAS 1303-96-4
CAS Max % 4
CAS 6381-59-5
CAS Max % 1
CAS 1313-84-4
CAS Max % 1
CAS 1310-73-2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC268401

 
lab chem
LC268401
Poly Bottle 500mL Each for $64.91

LC268402

 
lab chem
LC268402
Poly Bottle 1L Each for $87.26
Description & Specifications

Specifications

Chemical Name or Material Water Hardness Buffer
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 93.25
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May damage fertility or the unborn child.
Toxic to aquatic life.
CAS 1303-96-4
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 4
Recommended Storage Room Temperature
CAS 6381-59-5
CAS Max % 1
CAS 1313-84-4
CAS Max % 1
CAS 1310-73-2
CAS Max % 0.65
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Clear, almost colorless
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May damage fertility or the unborn child.
Toxic to aquatic life.

Room Temperature