missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water Hardness Buffer, Certified, LabChem™
Manufacturer: Lab Chem LC268004
Specifications
Water Hardness Buffer | |
141-43-5,7647-01-0,14402-88-1 | |
61.08 | |
Liquid | |
Odorless | |
Soluble in water | |
Yellow |
64,30,5.5,0.5 | |
C2H7NO | |
Passes Test | |
Certified | |
Poly Bottle | |
Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
4L |