missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Function tested and optimized for LC/MS applications

Supplier:  Avantor 983103

 View more versions of this product

Catalog No. 02004563



Description

Description

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Specifications

Specifications

Water
7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
BAKER ANALYZED™ LC-MS Reagent
4 L
Filtered through a 0.2 micron filter
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Narrow Mouth Amber Glass Bottle
1L = 1.00kg

Chemical Identifiers

7732-18-5
18.015
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
SDS
Documents

Documents

Promotions

Promotions