missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, EMD Millipore

$68.97 - $249.17

Chemical Identifiers

Molecular Formula H2O
Linear Formula H2O
Formula Weight 18.02
Synonym Aqua-Destillata
Chemical Name or Material LC-MS Water
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MWX00046 Emd Millipore Corporation
WX00046
Glass Bottle 1L Each for $68.97
MWX00036 Emd Millipore Corporation
WX00036
1L Each for $249.17
Specifications Description & Specifications

Chemical Identifiers

H2O
18.02
LC-MS Water
H2O
Aqua-Destillata

Specifications

18.02
H2O
Aqua-Destillata
LC-MS Water
1L
Glass Bottle
H2O