Please login to your online account to display your discounted pricing

Water, Deuterium-depleted, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Water
CAS 7732-18-5
Purity Grade For NMR
Molecular Formula H2O
Linear Formula H2O
Formula Weight 18.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187340250

  SDS
fisher scientific acros
187340250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 18.02
Physical Form Liquid
Quantity 25g
Color Clear colorless
Purity Grade For NMR
Density 1.0000g/mL
Merck Index 15, 10237
Fieser 16,383
CAS 7732-18-5
Molecular Formula H2O
Chemical Name or Material Water
Deuterium (D) 3ppm max.
Packaging Glass bottle
Linear Formula H2O

For NMR