Please login to your online account to display your discounted pricing

Water, Deuterium-depleted, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  187340250

 View more versions of this product

Catalog No. AC187340250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 18.02
Quantity 25g
Color Clear colorless
Density 1.0000g/mL
Purity Grade For NMR
Fieser 16,383
Merck Index 15, 10237
Chemical Name or Material Water
Molecular Formula H2O
CAS 7732-18-5
Deuterium (D) 3ppm max.
Packaging Glass bottle
Linear Formula H2O

Specifications

Chemical Name or Material Water
CAS 7732-18-5
Purity Grade For NMR
Molecular Formula H2O
Linear Formula H2O
Formula Weight 18.02