missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, Deionized, ASTM Type II, Certified, LabChem™
Supplier: LabChem LC26750200
Specifications
Water | |
7732-18-5 | |
0°C | |
100 | |
H2O | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Poly Drum | |
0.99823g/mL | |
0.99823g/mL |
Deionized, ASTM Type II | |
Colorless | |
100°C | |
<1uS/cm | |
H2O | |
Soluble in water | |
O | |
18.015 | |
CHEBI:15377 | |
Certified | |
1.004cSt | |
200 L | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling
GHS H Statement
Product is not hazardous.
EINECSNumber : 231-791-2
Recommended Storage : Room Temperature