missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water Blank For ICP, ASTM Type I, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$262.91 - $388.56

Specifications

Packaging Triple-leached LDPE bottle
Quantity 500 mL
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Assurance
Solvent or Matrix H2O
Chemical Name or Material Water Blank

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLBLK-H2O
View Documents Promotion Details
SPEX CertiPrep Group LLC
PLBLKH2O
500 mL Triple-leached LDPE bottle
Each for $262.91
 
PLBLKH2O-2L
View Documents Promotion Details
SPEX CertiPrep Group LLC
PLBLKH2O2L
2 L Triple-leached LDPE bottle
Each for $388.56
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

DI Water Matrix Blank

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Assurance
Water Blank
500 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
H2O
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only