missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, ACS Reagent, meets ASTM Type II specifications, Honeywell
$161.04 - $174.88
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7732-18-5,7732-18-5 | |
H2O | |
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified | |
O | |
18.015 | |
CHEBI:15377 | |
ACS Reagent | |
0.9971 g/cm3 | |
4 x 4L |
100.0000% | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18 | |
Poly Bottle | |
100°C | |
Water, ACS Reagent, meets ASTM Type II specifications |
Safety and Handling
ShelfLife : 547 days from date of manufacture