missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Water, ACS Reagent, Honeywell™ Riedel-de Haën™

ACS Reagent

$47.13 - $455.78

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046044
View Documents
Honeywell Riedel-de Haen™
320072500ML
500mL Poly Bottle
Each for $47.13
 
60046045
View Documents
Honeywell Riedel-de Haen™
3200726X500ML
6 x 500mL Poly Bottle
Case of 6 Each for $282.48
 
60046040
View Documents
Honeywell Riedel-de Haen™
32007225L
2.5L Poly Bottle
Each for $96.19
 
60046041
View Documents
Honeywell Riedel-de Haen™
3200724L
4L Poly Bottle
Each for $143.00
 
60046042
View Documents
Honeywell Riedel-de Haen™
3200724X2L
4 x 2L Fiberboard Box
Case of 4 Each for $259.84
 
60046043
View Documents
Honeywell Riedel-de Haen™
3200724X4L
4 x 4L Glass Bottle
Case of 4 Each for $455.78
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
CHEBI:15377
O

Specifications

7732-18-5
MFCD00011332
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02g/mol
Poly Bottle
1.000g/mL at 3.98°C
500mL
H2O
NONH for all modes of transport
dihydrogen monoxide, dihydrogen oxide, distilled water, purified water, sterile water, water, water vapor, water, deionized, water, mineral, water, purified
O
18.015
CHEBI:15377
ACS Reagent
0°C
100°C
Water
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

RTECSNumber : ZC0110000

SDS