missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, ACS Reagent, Honeywell Riedel-de Haën™
ACS Reagent
Supplier: Honeywell-Riedel-de Haen 3200724X4L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Water | |
0°C | |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
ACS Reagent | |
1.000g/mL at 3.98°C |
7732-18-5 | |
100°C | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
Glass Bottle | |
4 x 4 L |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000