missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
Supplier: Ricca Chemical Company 915025
Specifications
Water | |
7732-18-5 | |
0°C | |
100°C | |
max. 0.056 | |
dihydrogen oxide, dihydrogen monoxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.01 | |
Cubitainer | |
10 L |
ASTM Type I | |
Colorless | |
98.0 | |
100.0 | |
H2O | |
Miscible | |
O | |
18.015 | |
CHEBI:15377 | |
ACS | |
Non-sterile | |
Liquid |
Chemical Identifiers
7732-18-5 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
CHEBI:15377 |
H2O | |
962 | |
O |
Safety and Handling
ShelfLife : 18 months
DOTInformation : Not Regulated by DOT