missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wash Solution, For Surfactants, LabChem™

$76.51 - $76.51

Chemical Identifiers

CAS 7732-18-5,7558-80-7,7664-93-9
CAS Max % 94.97,4.35,0.68
Linear Formula NaH2PO4
Formula Weight 119.98
Chemical Name or Material Wash Solution, For Surfactants
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC267002 Lab Chem
LC267002
1L Each for $76.51
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7558-80-7,7664-93-9
NaH2PO4
Wash Solution, For Surfactants
94.97,4.35,0.68
119.98

Specifications

119.98
Passes Test
NaH2PO4
Phosphorus oxides; Sulfur compounds
1L
1g/mL
Soluble in water
Wash Solution, For Surfactants
7732-18-5,7664-93-9,7558-80-7
Room Temperature
0.68,4.35,94.97
Poly Bottle
Liquid
Clear, colorless
1g/mL

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.