Please login to your online account to display your discounted pricing

Alfa Aesar™ Wang resin, 1% cross-linked, 0.8-1.1mmol/g, 200-400 mesh

Chemical Identifiers

Chemical Name or Material Wang resin
Name Note 1% cross-linked
MDL Number MFCD00131778
Synonym 4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1702814

  SDS
alfa aesar™
L1702814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Wang resin
Synonym 4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
MDL Number MFCD00131778
Quantity 25g
Name Note 1% cross-linked
Concentration 0.8 to 1.1mmol/g
Mesh Size 200 to 400 mesh